6,7-Dimethoxyisobenzofuran-1(3H)-one 98%

Supplier: Ambeed
Warning

Synonyms: 6,7-Dimethoxy-3H-1-isobenzofuranone

A481234-100MG A481234-1G A481234-250MG
77017-026EA 13.8 USD
77017-026 77017-028 77017-030
6,7-Dimethoxyisobenzofuran-1(3H)-one 98%
6,7-Dimethoxyisobenzofuran-1(3H)-one

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MW: 194.184 g/mol
MDL Number: MFCD00219354
CAS Number: 569-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR