5-Bromo-6-(trifluoromethyl)pyrimidine-2,4(1H,3H)-dione 95+%
Supplier: Ambeed
|
Warning
|
A481193-100MG
A481193-1G
A481193-250MG
A481193-50MG
A481193-5G
77017-016EA
70
USD
77017-016
77017-018
77017-020
77017-022
77017-024
5-Bromo-6-(trifluoromethyl)pyrimidine-2,4(1H,3H)-dione 95+%
5-Bromo-6-(trifluoromethyl)pyrimidine-2,4(1H,3H)-dione
Precaución: for Research Use Only
Formula:
C₅H₂BrF₃N₂O₂ |
MDL Number:
MFCD20486240 CAS Number: 26676-21-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...