Methyl 2-amino-5-fluoroisonicotinate 95%

Supplier: Ambeed
Warning

A481071-100MG A481071-250MG
76991-006EA 12 USD
76991-006 76991-008
Methyl 2-amino-5-fluoroisonicotinate 95%
Methyl 2-amino-5-fluoroisonicotinate

Precaución: for Research Use Only

Formula: C₇H₇FN₂O₂
MDL Number: MFCD23161384
CAS Number: 1380331-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR