1-Nonanol 99%

Supplier: Ambeed
Warning

A480931-1000G A480931-100G A480931-5G A480931-25G A480931-500G
77004-582EA 53 USD
77004-582 77004-584 77004-586 77602-076 77602-078
1-Nonanol 99%
1-Nonanol

Precaución: for Research Use Only

Formula: H₃C(CH₂)₇CH₂OH
MDL Number: MFCD00002990
CAS Number: 143-08-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR