3-Hydroxy-1-methylpyridin-4(1H)-one 95%

Supplier: Ambeed
Warning

A480910-250MG A480910-1G A480910-100MG
76991-004EA 157.02 USD
76991-004 77594-502 77684-278
3-Hydroxy-1-methylpyridin-4(1H)-one 95%
3-Hydroxy-1-methylpyridin-4(1H)-one

Precaución: For research use only.

Formula: C₆H₇NO₂
MW: 125.13 g/mol
MDL Number: MFCD12405601
CAS Number: 50700-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR