Ethyl 5-bromo-1H-indazole-3-carboxylate 98%

Supplier: Ambeed
Warning

A480556-100MG A480556-250MG A480556-5G A480556-1G
77017-010EA 5 USD
77017-010 77017-012 77017-014 77890-972
Ethyl 5-bromo-1H-indazole-3-carboxylate 98%
Ethyl 5-bromo-1H-indazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂O₂
MDL Number: MFCD05663980
CAS Number: 1081-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR