Isoxazolidine hydrochloride 95%

Supplier: Ambeed
Warning

A480144-1G A480144-250MG A480144-5G A480144-100MG
77004-572EA 80 USD
77004-572 77004-574 77594-498 77882-208
Isoxazolidine hydrochloride 95%
Isoxazolidine hydrochloride

Precaución: for Research Use Only

Formula: C₃H₇NO·HCl
MDL Number: MFCD11100465
CAS Number: 39657-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR