2-Nitro-4,6-bis(trifluoromethyl)aniline 97%

Supplier: Ambeed
Warning

A480067-1G A480067-250MG A480067-25G A480067-5G
76990-988EA 21.22 USD
76990-988 76990-990 76990-992 77684-277
2-Nitro-4,6-bis(trifluoromethyl)aniline 97%
2-Nitro-4,6-bis(trifluoromethyl)aniline

Precaución: For research use only.

Formula: C₈H₄F₆N₂O₂
MW: 274.12 g/mol
MDL Number: MFCD25962254
CAS Number: 320-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR