1-(4-Bromophenyl)piperidine 98%

Supplier: Ambeed
Warning

A479686-250MG A479686-5G A479686-1G
77004-568EA 15.92 USD
77004-568 77004-570 77594-496
1-(4-Bromophenyl)piperidine 98%
1-(4-Bromophenyl)piperidine

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrN
MW: 240.14 g/mol
MDL Number: MFCD04112533
CAS Number: 22148-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR