1-tert-Butyl 2-methyl 4-oxopyrrolidine-1,2-dicarboxylate 98%

Supplier: Ambeed
Danger

A479465-1G A479465-250MG A479465-25G A479465-5G
77017-004EA 18.04 USD
77017-004 77017-006 77017-008 77613-500
1-tert-Butyl 2-methyl 4-oxopyrrolidine-1,2-dicarboxylate 98%
1-tert-Butyl 2-methyl 4-oxopyrrolidine-1,2-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₅
MW: 243.25 g/mol
MDL Number: MFCD03701320
CAS Number: 362706-26-1
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR