(4-Nitrophenyl)-β-D-galactopyranoside 98% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: pNPG, 4-Nitrophenyl β-D-Galactopyranoside

A479243-1G A479243-5G A479243-25G
77004-554EA 11 USD
77004-554 77602-070 77882-204
(4-Nitrophenyl)-β-D-galactopyranoside 98% (by HPLC)
(4-Nitrophenyl)-β-D-galactopyranoside

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₈
MDL Number: MFCD00063256
CAS Number: 3150-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR