2,4,4'-Trimethoxybenzophenone 95%

Supplier: Ambeed
Warning

Synonyms: (2,4-Dimethoxyphenyl)(4-methoxyphenyl)methanone

A478717-1G A478717-250MG
77034-328EA 47.75 USD
77034-328 77034-330
2,4,4'-Trimethoxybenzophenone 95%
2,4,4'-Trimethoxybenzophenone

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₄
MW: 272.30 g/mol
MDL Number: MFCD00100089
CAS Number: 4038-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR