2-Bromo-3-pyridineboronic acid 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A478311-100G A478311-1G A478311-25G A478311-5G
77017-000EA 251 USD
77017-000 77017-002 77684-274 77890-964
2-Bromo-3-pyridineboronic acid 97%, Ambeed.Inc
2-Bromo-3-pyridineboronic acid

Precaución: For research use only.

Formula: C₅H₅BBrNO₂
MDL Number: MFCD04113805
CAS Number: 452972-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR