2-(Trifluoromethoxy)phenylacetonitrile 98%

Supplier: Ambeed
Danger

A478028-1G A478028-250MG A478028-25G
77004-544EA 10 USD
77004-544 77004-546 77004-548
2-(Trifluoromethoxy)phenylacetonitrile 98%
2-(Trifluoromethoxy)phenylacetonitrile

Precaución: for Research Use Only

Formula: C₉H₆F₃NO
MDL Number: MFCD00236326
CAS Number: 137218-25-8
UN: 3276
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR