4-Chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide 98%

Supplier: Ambeed
Warning

A477769-100MG A477769-1G A477769-250MG
76990-970EA 39 USD
76990-970 76990-972 76990-974
4-Chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide 98%
4-Chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

Precaución: for Research Use Only

Formula: C₁₃H₁₇BClNO₃
MDL Number: MFCD18383526
CAS Number: 1242422-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR