2,6-Dibromopyridin-4-ol 95%
Supplier: Ambeed
|
Warning
|
A477759-10G
A477759-1G
A477759-250MG
A477759-25G
A477759-5G
76990-960EA
258.86
USD
76990-960
76990-962
76990-964
76990-966
76990-968
2,6-Dibromopyridin-4-ol 95%
2,6-Dibromopyridin-4-ol
Precaución: for Research Use Only
Formula:
C₅H₃Br₂NO MW: 252.89 g/mol |
MDL Number:
MFCD13185517 CAS Number: 220616-68-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...