tert-Butyl 3-amino-1H-pyrazole-1-carboxylate 97%

Supplier: Ambeed
Danger

A477362-1G A477362-250MG A477362-5G A477362-25G A477362-100MG
76990-954EA 23.34 USD
76990-954 76990-956 77572-500 77594-490 77684-271
tert-Butyl 3-amino-1H-pyrazole-1-carboxylate 97%
tert-Butyl 3-amino-1H-pyrazole-1-carboxylate

Precaución: For research use only.

Formula: C₈H₁₃N₃O₂
MW: 183.208 g/mol
MDL Number: MFCD13194910
CAS Number: 863504-84-1
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR