4-(4-Chlorophenyl)-2-isopropyl-2,8-diazaspiro[4.5]decan-1-one hydrochloride 98%

Supplier: Ambeed
Warning

A476768-100MG A476768-1G A476768-250MG
77034-322EA 36 USD
77034-322 77034-324 77034-326
4-(4-Chlorophenyl)-2-isopropyl-2,8-diazaspiro[4.5]decan-1-one hydrochloride 98%
4-(4-Chlorophenyl)-2-isopropyl-2,8-diazaspiro[4.5]decan-1-one hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₂₄Cl₂N₂O
MDL Number: MFCD08752144
CAS Number: 1198284-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR