2-Bromo-4-nitro-5-(trifluoromethyl)aniline 98%

Supplier: Ambeed
Warning

A476709-100MG A476709-1G A476709-250MG A476709-5G
77004-532EA 13.8 USD
77004-532 77004-534 77004-536 77004-538
2-Bromo-4-nitro-5-(trifluoromethyl)aniline 98%
2-Bromo-4-nitro-5-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₄BrF₃N₂O₂
MW: 285.0181 g/mol
MDL Number: MFCD12922693
CAS Number: 1190198-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR