N-Methyl-N-(2-hydroxyethyl)-4-aminobenzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 4-((2-Hydroxyethyl)(methyl)amino)benzaldehyde

A476620-1G A476620-5G
77004-528EA 19.1 USD
77004-528 77004-530
N-Methyl-N-(2-hydroxyethyl)-4-aminobenzaldehyde 98%
N-Methyl-N-(2-hydroxyethyl)-4-aminobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.216 g/mol
MDL Number: MFCD00963547
CAS Number: 1201-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR