Ethyl 8-(benzyloxy)-2-methylimidazo[1,2-a]pyridine-3-carboxylate 95%

Supplier: Ambeed
Warning

A475400-100MG A475400-1G A475400-250MG
77016-986EA 26.53 USD
77016-986 77016-988 77016-990
Ethyl 8-(benzyloxy)-2-methylimidazo[1,2-a]pyridine-3-carboxylate 95%
Ethyl 8-(benzyloxy)-2-methylimidazo[1,2-a]pyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₁₈N₂O₃
MW: 310.35 g/mol
MDL Number: MFCD06739215
CAS Number: 96428-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR