4-(3-Bromophenyl)-6,8-dichloro-2-methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride 98+%

Supplier: Ambeed
Warning

A474987-1G A474987-25G A474987-5G
77016-980EA 14.86 USD
77016-980 77016-982 77016-984
4-(3-Bromophenyl)-6,8-dichloro-2-methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride 98+%
4-(3-Bromophenyl)-6,8-dichloro-2-methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride

Precaución: for Research Use Only

Formula: C₁₆H₁₅BrCl₃N
MW: 407.56 g/mol
MDL Number: MFCD22683910
CAS Number: 543739-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR