4,5,9,10-Tetrabromoisochromeno[6,5,4-def]isochromene-1,3,6,8-tetraone 95%

Supplier: Ambeed
Warning

A474692-100MG A474692-1G A474692-250MG
77016-974EA 137.92 USD
77016-974 77016-976 77016-978
4,5,9,10-Tetrabromoisochromeno[6,5,4-def]isochromene-1,3,6,8-tetraone 95%
4,5,9,10-Tetrabromoisochromeno[6,5,4-def]isochromene-1,3,6,8-tetraone

Precaución: for Research Use Only

Formula: C₁₄Br₄O₆
MW: 583.76 g/mol
CAS Number: 299962-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR