(S)-2-Methylpiperidin-4-one hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A474502-1G
A474502-250MG
A474502-5G
77016-970EA
28
USD
77016-970
77016-972
77572-494
(S)-2-Methylpiperidin-4-one hydrochloride 98%
(S)-2-Methylpiperidin-4-one hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₁₂ClNO |
MDL Number:
MFCD22682260 CAS Number: 790667-45-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...