(4-((2-Methoxyethoxy)methyl)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

A474006-1G A474006-250MG A474006-5G
77016-960EA 188.99 USD
77016-960 77016-962 77016-964
(4-((2-Methoxyethoxy)methyl)phenyl)boronic acid 98%
(4-((2-Methoxyethoxy)methyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₄
MDL Number: MFCD14687278
CAS Number: 1146169-44-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR