2,8-Dichloroquinazolin-4-amine 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A473743-1G A473743-250MG A473743-5G
76990-946EA 46 USD
76990-946 76990-948 77684-269
2,8-Dichloroquinazolin-4-amine 97%, Ambeed.Inc
2,8-Dichloroquinazolin-4-amine

Precaución: For research use only.

Formula: C₈H₅Cl₂N₃
MDL Number: MFCD11858259
CAS Number: 1107694-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR