2-Hydroxy-N,N,N-trimethylethanaminium 2-(4-(4-chlorobenzoyl)phenoxy)-2-methylpropanoate 98%

Supplier: Ambeed
Warning

Synonyms: Choline Fenofibrate

A473645-100MG A473645-10MG A473645-1MG A473645-5G
76990-938EA 13.8 USD
76990-938 76990-940 76990-942 76990-944
2-Hydroxy-N,N,N-trimethylethanaminium 2-(4-(4-chlorobenzoyl)phenoxy)-2-methylpropanoate 98%
2-Hydroxy-N,N,N-trimethylethanaminium 2-(4-(4-chlorobenzoyl)phenoxy)-2-methylpropanoate

Precaución: for Research Use Only

Formula: C₂₂H₂₈ClNO₅
MW: 421.91 g/mol
MDL Number: MFCD18382233
CAS Number: 856676-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR