(2-(Diphenylphosphino)phenyl)methanamine 98%
Supplier: Ambeed
|
Warning
|
A473369-100MG
A473369-1G
A473369-250MG
77016-940EA
84
USD
77016-940
77016-942
77016-944
(2-(Diphenylphosphino)phenyl)methanamine 98%
(2-(Diphenylphosphino)phenyl)methanamine
Precaución: for Research Use Only
Formula:
C₁₉H₁₈NP |
CAS Number:
177263-77-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...