3,5-Dihydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-en-1-yl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one 95%

Supplier: Ambeed
Warning

Synonyms: Icariside I

A473088-10MG A473088-1MG A473088-25MG A473088-5MG
76990-930EA 39.26 USD
76990-930 76990-932 76990-934 76990-936
3,5-Dihydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-en-1-yl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one 95%
3,5-Dihydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-en-1-yl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₂₇H₃₀O₁₁
MW: 530.52 g/mol
MDL Number: MFCD01662755
CAS Number: 56725-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR