3,4-Dihydro-3-oxo-2H-1,4-benzoxazine-6-carbonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 3-Oxo-3,4-dihydro-2H-benzo[b][1,4]oxazine-6-carbonitrile, 3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carbonitrile

A472729-100MG A472729-1G A472729-250MG
76990-918EA 43.5 USD
76990-918 76990-920 76990-922
3,4-Dihydro-3-oxo-2H-1,4-benzoxazine-6-carbonitrile 95%
3,4-Dihydro-3-oxo-2H-1,4-benzoxazine-6-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₆N₂O₂
MW: 174.156 g/mol
MDL Number: MFCD20039908
CAS Number: 134997-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR