5-(Boc-amino)pyridine-3-boronic acid pinacol ester 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl (5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl)carbamate
A472367-100MG
A472367-1G
A472367-250MG
A472367-5G
77034-284EA
30
USD
77034-284
77034-286
77034-288
77034-290
5-(Boc-amino)pyridine-3-boronic acid pinacol ester 95%
5-(Boc-amino)pyridine-3-boronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₆H₂₅BN₂O₄ |
MDL Number:
MFCD13195277 CAS Number: 1171897-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...