tert-Butyl 3-methyl-1H-pyrazolo[3,4-b]pyridine-1-carboxylate 95+%

Supplier: Ambeed
Warning

A472272-100MG A472272-250MG
77016-934EA 107.16 USD
77016-934 77016-936
tert-Butyl 3-methyl-1H-pyrazolo[3,4-b]pyridine-1-carboxylate 95+%
tert-Butyl 3-methyl-1H-pyrazolo[3,4-b]pyridine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₅N₃O₂
MW: 233.26 g/mol
MDL Number: MFCD11616024
CAS Number: 174180-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR