2-Ethoxy-5-nitropyridine 97%

Supplier: Ambeed
Warning

A471808-100G A471808-5G
77004-480EA 225.98 USD
77004-480 77004-482
2-Ethoxy-5-nitropyridine 97%
2-Ethoxy-5-nitropyridine

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
MW: 168.15 g/mol
MDL Number: MFCD01646181
CAS Number: 31594-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR