5-Bromo-3-chloro-2-hydroxybenzonitrile 95%

Supplier: Ambeed
Warning

A471450-100MG A471450-1G
76990-914EA 57 USD
76990-914 76990-916
5-Bromo-3-chloro-2-hydroxybenzonitrile 95%
5-Bromo-3-chloro-2-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃BrClNO
MDL Number: MFCD16249641
CAS Number: 1260810-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR