6-Methoxyindole-2-boronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxy-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

A471433-1G A471433-250MG A471433-5G
77016-924EA 208 USD
77016-924 77016-926 77016-928
6-Methoxyindole-2-boronic acid pinacol ester 97%
6-Methoxyindole-2-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₀BNO₃
MDL Number: MFCD16295111
CAS Number: 1256359-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR