(1-Hydroxy-2-(pyridin-3-yl)ethane-1,1-diyl)diphosphonic acid 99% (by HPLC)

Supplier: Ambeed
Warning

A471285-100G A471285-1G A471285-25G A471285-5G
77004-472EA 216.43 USD
77004-472 77004-474 77004-476 77004-478
(1-Hydroxy-2-(pyridin-3-yl)ethane-1,1-diyl)diphosphonic acid 99% (by HPLC)
(1-Hydroxy-2-(pyridin-3-yl)ethane-1,1-diyl)diphosphonic acid

Precaución: for Research Use Only

Formula: C₇H₁₁NO₇P₂
MW: 283.11 g/mol
MDL Number: MFCD00867080
CAS Number: 105462-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR