1,6-Dibromonaphthalene 95%

Supplier: Ambeed
Warning

A471218-100MG A471218-1G A471218-250MG
77034-278EA 18 USD
77034-278 77034-280 77034-282
1,6-Dibromonaphthalene 95%
1,6-Dibromonaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₆Br₂
MDL Number: MFCD16620284
CAS Number: 19125-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR