(1R,2R)-N1,N2-Bis(2-(bis(3,5-dimethylphenyl)phosphino)benzyl)cyclohexane-1,2-diamine ≥98%, ee 99%

Supplier: Ambeed
Warning

A471203-100MG A471203-1G A471203-250MG
76990-908EA 54 USD
76990-908 76990-910 76990-912
(1R,2R)-N1,N2-Bis(2-(bis(3,5-dimethylphenyl)phosphino)benzyl)cyclohexane-1,2-diamine ≥98%, ee 99%
(1R,2R)-N1,N2-Bis(2-(bis(3,5-dimethylphenyl)phosphino)benzyl)cyclohexane-1,2-diamine

Precaución: for Research Use Only

Formula: C₅₂H₆₀N₂P₂
MW: 774.99 g/mol
MDL Number: MFCD17014020
CAS Number: 1150113-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR