5-Chloroquinolin-4-ol 95%

Supplier: Ambeed
Warning

A470632-100MG A470632-1G A470632-50MG
76990-902EA 25 USD
76990-902 76990-904 76990-906
5-Chloroquinolin-4-ol 95%
5-Chloroquinolin-4-ol

Precaución: for Research Use Only

Formula: C₉H₆ClNO
MDL Number: MFCD16659125
CAS Number: 23443-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR