5-Chloroquinoxalin-2-ol 95%

Supplier: Ambeed
Warning

A470159-1G A470159-250MG A470159-5G
77016-912EA 22 USD
77016-912 77016-914 77016-916
5-Chloroquinoxalin-2-ol 95%
5-Chloroquinoxalin-2-ol

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O
MDL Number: MFCD11846472
CAS Number: 55687-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR