N-Benzyloxycarbonyl-3-fluoro-4-morpholinoaniline 98%

Supplier: Ambeed
Warning

Synonyms: N-benzyloxycarbonyl-3-fluoro-4-morpholinoaniline, (3-Fluoro-4-morpholinophenyl)carbamic acid Benzyl ester, Benzyl (3-Fluoro-4-morpholinophenyl)carbamate

A469625-25G A469625-5G A469625-100G A469625-500G
77034-268EA 14 USD
77034-268 77034-270 77673-206 77872-781
N-Benzyloxycarbonyl-3-fluoro-4-morpholinoaniline 98%
N-Benzyloxycarbonyl-3-fluoro-4-morpholinoaniline

Precaución: for Research Use Only

Formula: C₁₈H₁₉FN₂O₃
MW: 330.359 g/mol
MDL Number: MFCD06658240
CAS Number: 168828-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR