4-Octylphenol 99%

Supplier: Ambeed
Warning

A468192-1G A468192-250MG A468192-5G
77004-458EA 19.1 USD
77004-458 77004-460 77004-462
4-Octylphenol 99%
4-Octylphenol

Precaución: for Research Use Only

Formula: C₁₄H₂₂O
MW: 206.33 g/mol
MDL Number: MFCD00036134
CAS Number: 1806-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR