tert-Butyl 4-(hydroxyimino)piperidine-1-carboxylate 98%

Supplier: Ambeed
Danger

A467950-10G A467950-1G A467950-5G
77034-262EA 65.78 USD
77034-262 77034-264 77034-266
tert-Butyl 4-(hydroxyimino)piperidine-1-carboxylate 98%
tert-Butyl 4-(hydroxyimino)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₃
MW: 214.26 g/mol
MDL Number: MFCD09953970
CAS Number: 150008-24-5
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR