4,4'-Bis(chloromethyl)biphenyl 95%

Supplier: Ambeed
Danger

A467732-10G A467732-25G
77004-454EA 5 USD
77004-454 77551-422
4,4'-Bis(chloromethyl)biphenyl 95%
4,4'-Bis(chloromethyl)biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂Cl₂
MDL Number: MFCD00674019
CAS Number: 1667-10-3
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR