(4-((3-(Trifluoromethyl)phenoxy)methyl)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

A467138-1G A467138-250MG
77016-892EA 84.88 USD
77016-892 77016-894
(4-((3-(Trifluoromethyl)phenoxy)methyl)phenyl)boronic acid 98%
(4-((3-(Trifluoromethyl)phenoxy)methyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂BF₃O₃
MW: 296.05 g/mol
MDL Number: MFCD07369740
CAS Number: 849062-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR