Methyl (R)-(+)-3-boc-2,2-dimethyl-4-oxazolidinecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: (R)-(+)-3-Boc-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, (R)-(+)-3-(tert-Butoxycarbonyl)-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, Methyl (R)-N-Boc-2,2-dimethyloxazolidine-4-carboxylate, (R)-(+)-2,2-Dimethyloxazolidine-3,4-dicarboxylic acid 3-tert-Butyl 4-Methyl ester, 3-tert-Butyl 4-Methyl (R)-(+)-2,2-Dimethyloxazolidine-3,4-dicarboxylate

A466943-100G A466943-1G A466943-250MG A466943-25G A466943-5G
76990-892EA 580.32 USD
76990-892 76990-894 76990-896 76990-898 77613-436
Methyl (R)-(+)-3-boc-2,2-dimethyl-4-oxazolidinecarboxylate 97%
Methyl (R)-(+)-3-boc-2,2-dimethyl-4-oxazolidinecarboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₅
MW: 259.302 g/mol
MDL Number: MFCD00674041
CAS Number: 95715-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR