(1S,2S)-N1,N2-Dimethyl-1,2-bis(3-(trifluoromethyl)phenyl)ethane-1,2-diamine 98%
Supplier: Ambeed
|
Warning
|
A466709-100MG
A466709-250MG
A466709-1G
76990-888EA
98.67
USD
76990-888
76990-890
77684-256
(1S,2S)-N1,N2-Dimethyl-1,2-bis(3-(trifluoromethyl)phenyl)ethane-1,2-diamine 98%
(1S,2S)-N1,N2-Dimethyl-1,2-bis(3-(trifluoromethyl)phenyl)ethane-1,2-diamine
Precaución: For research use only.
Formula:
C₁₈H₁₈F₆N₂ MW: 376.33 g/mol |
MDL Number:
MFCD00075561 CAS Number: 205873-26-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...