3-Iodoquinoline 95%

Supplier: Ambeed
Warning

A466413-100MG A466413-250MG A466413-5G A466413-1G
76990-876EA 112.46 USD
76990-876 76990-878 76990-880 77684-255
3-Iodoquinoline 95%
3-Iodoquinoline

Precaución: For research use only.

Formula: C₉H₆IN
MW: 255.05 g/mol
MDL Number: MFCD11656390
CAS Number: 79476-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR