(S)-1-(5-Fluoro-2-methylphenyl)ethanamine hydrochloride 95%

Supplier: Ambeed
Warning

A466370-100MG A466370-1G A466370-250MG
76990-870EA 64 USD
76990-870 76990-872 76990-874
(S)-1-(5-Fluoro-2-methylphenyl)ethanamine hydrochloride 95%
(S)-1-(5-Fluoro-2-methylphenyl)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₃ClFN
MDL Number: MFCD16294101
CAS Number: 2437254-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR