3-(2-Hydroxyethyl)benzoic acid 95%

Supplier: Ambeed
Warning

A466030-250MG A466030-1G
76990-868EA 66 USD
76990-868 77572-466
3-(2-Hydroxyethyl)benzoic acid 95%
3-(2-Hydroxyethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MDL Number: MFCD09027950
CAS Number: 320730-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR